missing translation for 'onlineSavingsMsg'
Learn More

Trans-4-(Boc-aminomethyl)cyclohexanmethanol, 97%, Thermo Scientific Chemicals

Catalog Number p-7041606
Click to view available options
Menge:
250 mg
1 g
5 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
172348-63-9
C13H25NO3
243.347
MFCD06657666
KSXPGASLEFGROT-UHFFFAOYSA-N
tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
18397396
tert-Butyl-N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamat
CC(C)(C)OC(=O)NCC1CCC(CC1)CO
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
172348-63-9
C13H25NO3
243.347
MFCD06657666
KSXPGASLEFGROT-UHFFFAOYSA-N
tert-butyl trans-4-hydroxymethylcyclohexylmethyl-carbamate, trans-4-boc-aminomethyl-1-hydroxymethyl-cyclohexane, tert-butyl trans-4-hydroxymethyl cyclohexyl methyl carbamate, tert-butyl trans-4-hydroxymethylcyclohexylmethyl carbamate, tert-butyl 4-hydroxymethyl cyclohexyl methylcarbamate, ksxpgaslefgrot-xypyzodxsa-n, tert-butyl 4-hydroxymethyl cyclohexyl methyl carbamate, trans-4-n-tert-butoxycarbonylaminomethyl cyclohexylmethanol, n-tert-butoxycarbonyl-trans-4-aminomethyl cyclohexanemethanol
18397396
tert-Butyl-N-[[4-(hydroxymethyl)cyclohexyl]methyl]carbamat
CC(C)(C)OC(=O)NCC1CCC(CC1)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material trans-4-(Boc-aminomethyl)cyclohexanemethanol
Formelmasse 243.34
Reinheit (%) 97%
Menge 250 mg

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'