missing translation for 'onlineSavingsMsg'
Learn More

trans-Clopidogrel-MP-13C,d3 Ethyl Ester Derivative(Mixture of Diastereomers), TRC

Product Code. 30367245
Change view
Click to view available options
Menge:
1 mg
5 mg
10 mg
Unit Size:
1 Milligramm
10 Milligramm
5 Milligramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
30367245 10 mg 10 Milligramm
30365520 5 mg 5 Milligramm
30362875 1 mg 1 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30367245 Supplier Toronto Research Chemicals Supplier No. TRCC58727710MG

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1331383-23-3
Summenformel C2613CH27D3ClNO6S
Molekulargewicht (g/mol) 536.06
Synonym Trans-Clopidogrel-Mp-13C,D3 Ethyl Ester Derivative, (Mixture Of Diastereomers)
IUPAC-Name methyl 2-(2-chlorophenyl)-2-[(3E)-3-(2-ethoxy-2-oxoethylidene)-4-[2-oxo-2-[3-(trideuterio(113C)methoxy)phenyl]ethyl]sulfanylpiperidin-1-yl]acetate
SMILES [2H][13C]([2H])([2H])Oc1cccc(c1)C(=O)CSC2CCN(C/C/2=C\C(=O)OCC)C(C(=O)OC)c3ccccc3Cl

Specifications

Chemischer Name oder Material trans-Clopidogrel-MP-13C,d3 Ethyl Ester Derivative(Mixture of Diastereomers)
Menge 10 mg
InChI-Formel InChI=1S/C27H30ClNO6S/c1-4-35-25(31)15-19-16-29(26(27(32)34-3)21-10-5-6-11-22(21)28)13-12-24(19)36-17-23(30)18-8-7-9-20(14-18)33-2/h5-11,14-15,24,26H,4,12-13,16-17H2,1-3H3/b19-15+/i2+1D3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.