missing translation for 'onlineSavingsMsg'
Learn More

Trans-N-Boc-4-Cyano-L-Proline Methyl-Ester, 97 %, Thermo Scientific Chemicals

Catalog Number p-7035031
Click to view available options
Menge:
250 mg
1 g
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
194163-91-2
C12H18N2O4
254.29
MFCD03094915
GHLKJIQVORAVHE-UHFFFAOYNA-N
boc-trans-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, n-boc-trans-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-cyanopyrrolidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-trans-4-cyano-l-proline methyl ester, 4r-1-tert-butyloxycarbonyl-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r
17998966
1-O-tert-Butyl-2-O-methyl-(2S,4R)-4-cyanopyrrolidin-1,2-dicarboxylat
COC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
194163-91-2
C12H18N2O4
254.29
MFCD03094915
GHLKJIQVORAVHE-UHFFFAOYNA-N
boc-trans-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butyl 2-methyl 4-cyanopyrrolidine-1,2-dicarboxylate, n-boc-trans-4-cyano-l-proline methyl ester, 1-tert-butyl 2-methyl 2s,4r-4-cyanopyrrolidine-1,2-dicarboxylate, n-tert-butoxycarbonyl-trans-4-cyano-l-proline methyl ester, 4r-1-tert-butyloxycarbonyl-4-cyano-l-proline methyl ester, 2s,4r-1-tert-butoxycarbonyl-4-cyano-2-methylpyrrolidine-2-carboxylic acid, 1,2-pyrrolidinedicarboxylicacid,4-cyano-,1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylic acid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r, 1,2-pyrrolidinedicarboxylicacid, 4-cyano-, 1-1,1-dimethylethyl 2-methyl ester, 2s,4r
17998966
1-O-tert-Butyl-2-O-methyl-(2S,4R)-4-cyanopyrrolidin-1,2-dicarboxylat
COC(=O)C1CC(CN1C(=O)OC(C)(C)C)C#N

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Chemischer Name oder Material trans-N-Boc-4-cyano-L-proline methyl ester
Menge 250 mg
Formelmasse 254.29
Reinheit (%) 97%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'