Learn More
trans-Stilbene-alpha,alpha'-d2, CDN
Specifications
Specifications
| Chemischer Name oder Material | trans-Stilbene-alpha,alpha'-D2 |
| CAS | 5284-44-6 |
| Summenformel | C14 D2 H10 |
| Menge | 0.1 g |
| Synonym | Benzene, 1,1'-[(1 E)-1,2-ethenediyl-1,2-d2]bis- (9 CI, ACI), 1,1'-[(1 E)-1,2-Ethenediyl-1,2-d2]bis[benzene] (ACI), Benzene, 1,1'-(1,2-ethenediyl-1,2-d2)bis-, (E)- (ZCI), Stilbene-α,α'-d2, (E)- (8 CI), trans-Stilbene-α,α'-d2, trans-Stilbene-α,α'-D2 |
| InChI-Formel | InChI=1 S/C14H12/c1-3-7-13(8-4-1)11-12-14-9-5-2-6-10-14/h1-12 H/b12-11+/i11D,12 D |
| SMILES | [2 H]\C(=C(\[2 H])/c1ccccc1)\c2ccccc2 |
| IUPAC-Name | [(E)-1,2-Dideuterio-2-phenylethenyl]benzol |
| Molekulargewicht (g/mol) | 182.257 |
| Formelmasse | 182.106 g/mol |
| Show More |