missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tri(Propylenglykolbutylether (Isomerengemisch), TRC)
Specifications
Specifications
| Chemischer Name oder Material | 1-[1-(1-butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol |
| CAS | 55934-93-5 |
| Summenformel | C13 H28 O4 |
| Menge | 10 mL |
| Synonym | Tri(Propylene Glycol Butyl Ether (Mixture of isomers), [2-(2-Butoxymethylethoxy)methylethoxy]propanol, 2-(2-Butoxymethylethoxy)methylethoxy)propanol, BFTG, Dalpad C, Dowanol TPnB, TPnB, Tripropylene Glycol Monobutyl Ether, Tripropylene Glycol n-Butyl Ether, Tripropyleneglycol Butyl Ether |
| InChI-Formel | InChI=1S/C13H28O4/c1-5-6-7-15-9-12(3)17-10-13(4)16-8-11(2)14/h11-14H,5-10H2,1-4H3 |
| SMILES | CCCCOCC(C)OCC(C)OCC(C)O |
| IUPAC-Name | 1-[1-(1-Butoxypropan-2-yloxy)propan-2-yloxy]propan-2-ol |
| Molekulargewicht (g/mol) | 248.359 |
| Formelmasse | 248.199 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?