Learn More
(Triethylsilyl)acetylene, 97%
Product Code. 11359066
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359066 5 g 5g
11349066 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11359066 Supplier Thermo Scientific Alfa Aesar Supplier No. L20217.06
 Restricted Item
Verfügbar ab: 21-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 1777-03-3 | C8H16Si | 140.30 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1777-03-3
Molecular Formula C8H16Si
Molecular Weight (g/mol) 140.30
MDL Number MFCD00075373
InChI Key FWSPXZXVNVQHIF-UHFFFAOYSA-N
Synonym triethylsilyl acetylene, ethynyltriethylsilane, triethyl ethynyl silane, triethylsilylacetylene, silane, triethylethynyl, triethylsilyl acetylen, triethylsilyl-acetylene, triethyl-ethynyl-silane
PubChem CID 637947
IUPAC Name triethyl(ethynyl)silane
SMILES CC[Si](CC)(CC)C#C
Specifications
Boiling Point 136°C to 140°C
Quantity 5 g
Flash Point 17°C (62°F)
Refractive Index 1.4315
Density 0.783
UN Number UN1993
Beilstein 1743814
Formula Weight 140.31
Percent Purity 97%
Chemical Name or Material (Triethylsilyl)acetylene
compliance-icons compliance-icons
Product Identifier
  • (Triethylsilyl)acetylene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: (Triethylsilyl)acetylene