Learn More
Trifluoroacetic Acid D1 99.5% D, Eurisotop
Product Code. 17257396
Encompass
Change view
Click to view available options
Quantity:
10 mL
2 x 0.75 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
17257396 10 mL
17247396 2 x 0.75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17257396 Supplier Eurisotop Supplier No. D022EA
 Restricted Item
Verfügbar ab: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Synonym: Trifluoroacetic Acid-d1
Trifluoroacetic Acid D1
  • Isotopic Enrichment: 99.50% D
CoA and MSDS available at Eurisotop.com
Chemical Identifiers
CAS 599-00-8
Molecular Formula C2HF3O2
Molecular Weight (g/mol) 115.03
InChI Key DTQVDTLACAAQTR-DYCDLGHISA-N
Synonym Trifluoroacetic acid-D1
IUPAC Name trifluoroethan(²H)oic acid
SMILES [2H]OC(=O)C(F)(F)F
Specifications
Color Colorless
Deuterium (D) 99.5% D
Quantity 10 mL
Melting Point -15.4°C
Boiling Point 72.4°C
Grade Research Grade
Chemical Name or Material Trifluoroacetic Acid D1