Learn More
Trimethyl(bromodifluoromethyl)silane, TRC
Product Code. 30456247
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456247 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30456247 Supplier Toronto Research Chemicals Supplier No. TRCT895825100MG
 Restricted Item
Verfügbar ab: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Trimethyl(bromodifluoromethyl)silane
CAS 115262-01-6
Molecular Formula C4H9F2SiBr
Quantity 100 mg
Synonym (Bromodifluoromethyl)trimethylsilane
InChI Formula InChI=1S/C4H9BrF2Si/c1-8(2,3)4(5,6)7/h1-3H3
SMILES C[Si](C)(C)C(Br)(F)F
Molecular Weight (g/mol) 203.1