Learn More
TRIS-acetate-SDS running buffer (10X), pH 8.3, Thermo Scientific™
Click to view available options
Quantity:
500 mL
1 L
Unit Size:
1 L
500mL
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
| Chemical Name or Material | TRIS-acetate-SDS running buffer (10X) |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | Plastic bottle |