missing translation for 'onlineSavingsMsg'
Learn More

Tris DBA, Tocris Bioscience™

N-myristoyltransferase-1 inhibitor; antiproliferative

Marke:  Tocris (Bio-Techne) 341650

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13412984

  • 555.12 € / 50 Milligramm


Voraussichtlicher Liefertermin 24-04-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

N-myristoyltransferase-1 (NMT-1) inhibitor that prevents activation of MAPK, PI 3-K and STAT3 signaling pathways. Exhibits antiproliferative activity against melanoma cells in vitro and in vivo. Also used as a cyclization catalyst.
Spezifikation

Chemische Identifikatoren

51364-51-3
915.734
tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium, tris dibenzylideneacetone dipalladium 0
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
C51H42O3Pd2
CYPYTURSJDMMMP-WVCUSYJESA-N
9811564
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.[Pd].[Pd]

Spezifikation

Tris DBA
51364-51-3
tris dibenzylideneacetone dipalladium 0, tris dibenzylideneacetone dipalladium, pd2 dba 3, tris dibezylideneacetone dipalladium, tris dibenzylideneacetone dipalladium, tris dibenzylideneacetone dipalladium o, tris dibenzylideneacetonyl bis-palladium, tris dba, tris 1e,4e-1,5-diphenylpenta-1,4-dien-3-one dipalladium, tris dibenzylideneacetone dipalladium 0
C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.C1=CC=C(C=C1)C=CC(=O)C=CC2=CC=CC=C2.[Pd].[Pd]
915.734
915.73
50mg
C51H42O3Pd2
CYPYTURSJDMMMP-WVCUSYJESA-N
(1E,4E)-1,5-diphenylpenta-1,4-dien-3-one;palladium
9811564
>98%
Videos