Learn More
Tromethamine, TRC
Artikelnummer. 30138089
Change view
Click to view available options
Menge:
50 g
100 g
250 g
Packungsgröße:
100 Gramm
250 Gramm
50 Gramm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
30138089 250 g 250 Gramm
17949834 100 g 100 Gramm
17919844 50 g 50 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138089 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCT892600250G
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 77-86-1
Summenformel C4 H11 N O3
Molekulargewicht (g/mol) 121.14
Synonym Trometamol, aminomethylidynetri(methanol), 1,3-Propanediol, 2-amino-2-(hydroxymethyl)-, 2-Amino-1,3-dihydroxy-2-(hydroxymethyl)propane, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-hydroxymethylpropan-1,3-diol, 2-Amino-2-methylol-1,3-propanediol, Addex-Tham, Aminotri(hydroxymethyl)methane, Aminotrimethylolmethane, Aminotris(hydroxymethyl)methane, Methanamine, 1,1,1-tris(hydroxymethyl)-, NSC 103026, NSC 6365, NSC 65434, Pehanorm, TAM, TAM (buffering agent), THAM, Talatrol, Tri Amino, Tri(hydroxymethyl)aminomethane, Tri(hydroxymethyl)methylamine, Trigmo base, Triladyl, Trimethylolaminomethane, Tris, Tris (buffering agent), Tris Amino, Tris Amino Crystal, Tris base, Tris buffer, Tris(hydroxymethyl)aminomethane, Tris(hydroxymethyl)methanamine, Tris(hydroxymethyl)methylamine, Tris(methylolamino)methane, Tris-steril, Trisamin, Trisamine, Trisaminol, Trizma, Trizma Base, Trometamole, Tromethamine, Tromethane, Tromethanmin, Tutofusin tris, [2-Hydroxy-1,1-bis(hydroxymethyl)ethyl]amine
IUPAC-Name 2-amino-2-(hydroxymethyl)propane-1,3-diol
SMILES NC(CO)(CO)CO
Spezifikation
Chemischer Name oder Material Trometamol
Hinweise zur Reinheitsqualität HPLC
Menge 250 g
InChI-Formel InChI=1S/C4H11NO3/c5-4(1-6,2-7)3-8/h6-8H,1-3,5H2
Formelmasse 121.0739
Reinheit (%) >95%