missing translation for 'onlineSavingsMsg'
Learn More

Troponin I Type 3 (cardiac) Antibody (1E10) - Azide and BSA Free, Novus Biologicals™

Product Code. 18324730 Shop All Bio Techne Products
Change view
Click to view available options
Menge:
0.1 mg
Unit Size:
0.10 Milligramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
18324730 0.1 mg 0.10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18324730 Supplier Novus Biologicals Supplier No. NBP280996

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Troponin I Type 3 (cardiac) Monoclonal specifically detects Troponin I Type 3 (cardiac) in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Troponin I Type 3 (cardiac)
Anwendungen Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry
Klassifikation Monoclonal
Klon 10000000000
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot : 0.5 μg/mL, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin : 5 μg/mL, Immunocytochemistry
Zusammensetzung PBS
Gen-Alias Cardiac troponin I, CMD1FF, CMD2A, CMH7MGC116817, cTnI, familial hypertrophic cardiomyopathy 7, RCM1, TNNC1troponin I, cardiac, troponin I type 3 (cardiac), troponin I, cardiac muscle
Gensymbole TNNI3
Wirtsspezies Mouse
Immunogen Partial recominant human Troponin 1 (between amino acids 1-200) [UniProt P19429]
Reinigungsverfahren Protein G purified
Menge 0.1 mg
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Cell Biology, Cellular Markers, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity, Phospho Specific, Signal Transduction, Stem Cell Markers
Primär oder sekundär Primary
Gen-ID (Entrez) 7137
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.