missing translation for 'onlineSavingsMsg'
Learn More

Trovafloxacin mesylate, Tocris Bioscience™

Antibiotic; inhibits bacterial DNA synthesis

Marke:  Tocris (Bio-Techne) 386350

Weitere Details : Netto-Gewicht : 0.01000kg

Menge. 50mg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 13413724

  • 916.43 € / 50 Milligramm


Voraussichtlicher Liefertermin 20-02-2019
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Fluoroquinolone antibiotic. Inhibits bacterial DNA topoisomerase IV and DNA gyrase and forms a stable quinolone-DNA complex with these enzymes which reversibly inhibits DNA synthesis. Displays potent activity against gram-positive and gram-negative bacteria. Increases the production of mitochondrial NO in immortalized hepatocytes; also increases mitochondrial Ca2+. Inhibits Panx-1 (IC50 ∽ 4μM).
Spezifikation

Chemische Identifikatoren

147059-75-4
512.46
trovafloxacin mesylate, trovan, trovafloxacin monomethanesulfonate, trovafloxacin methanesulfonate, unii-0p1lko80wn, 0p1lko80wn, trovafloxacin mesylate usan, trovan tn, trovafloxacin mesilate, dsstox_cid_25701
7-[(1S,5R)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
C21H19F3N4O6S
DYNZICQDCVYXFW-GIPYJWDTSA-N
62960
CS(=O)(=O)O.C1C2C(C2N)CN1C3=C(C=C4C(=O)C(=CN(C4=N3)C5=C(C=C(C=C5)F)F)C(=O)O)F

Spezifikation

Trovafloxacin mesylate
147059-75-4
trovafloxacin mesylate, trovan, trovafloxacin monomethanesulfonate, trovafloxacin methanesulfonate, unii-0p1lko80wn, 0p1lko80wn, trovafloxacin mesylate usan, trovan tn, trovafloxacin mesilate, dsstox_cid_25701
CS(=O)(=O)O.C1C2C(C2N)CN1C3=C(C=C4C(=O)C(=CN(C4=N3)C5=C(C=C(C=C5)F)F)C(=O)O)F
512.46
512.46
50mg
C21H19F3N4O6S
DYNZICQDCVYXFW-GIPYJWDTSA-N
7-[(1S,5R)-6-amino-3-azabicyclo[3.1.0]hexan-3-yl]-1-(2,4-difluorophenyl)-6-fluoro-4-oxo-1,8-naphthyridine-3-carboxylic acid;methanesulfonic acid
62960
>99%
Videos