missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA10IP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548194
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TSGA10IP Polyclonal antibody specifically detects TSGA10IP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TSGA10IP | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q3SY00 | |
TSGA10IP | |
KLH conjugated synthetic peptide between 492-520 amino acids from the C-terminal region of human TSGA10IP. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
TSGA10IP | |
FAM161C; testis specific 10 interacting protein; testis specific, 10 interacting protein; testis-specific protein 10-interacting protein; Tsga10-interacting protein; TSGA10IP | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
254187 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |