Learn More
TSGA2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553601
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: TAELIHLNHR YQGKFLNKNP VGPGKYVFDV GCEQHGEYRL TDMERGEEEE EEELVTVVPK WKATQITELA LWTPTL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 90%.
TSGA2 Polyclonal antibody specifically detects TSGA2 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TSGA2 | |
Polyclonal | |
Unconjugated | |
RSPH1 | |
ABCB2; cancer/testis antigen 79; CT79; h-meichroacidin; Male meiotic metaphase chromosome-associated acidic protein; MCA; Meichroacidin; MGC126568; MGC141927; PSF1; radial spoke head 1 homolog; radial spoke head 1 homolog (Chlamydomonas); RING4; RSP44; Rsph1; RSPH10A; testes specific gene A2 homolog; testis specific A2 homolog; testis specific gene A2; testis-specific gene A2 protein; TSA2; TSGA2; TSGA2 epididymal isoform | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
89765 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WYR4 | |
RSPH1 | |
Recombinant protein corresponding to Human TSGA2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only