Learn More
TSPAN13 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA552434
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ACLALNQEQQ GQLLEVGWNN TASARNDIQR NLNCCGFRSV NPNDTCLASC VKSDHSCSPC APIIGEYAGE V Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 87%.
TSPAN13 Polyclonal antibody specifically detects TSPAN13 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TSPAN13 | |
Polyclonal | |
Unconjugated | |
TSPAN13 | |
1100001I23Rik; NET6; NET-6; tetraspan NET-6; Tetraspan Tspan-13; tetraspanin 13; tetraspanin-13; Tm4sf13; transmembrane 4 superfamily member 13; transmembrane 4 superfamily member tetraspan NET-6; Tspan13; tspan-13; UNQ260/PRO296 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
27075 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95857 | |
TSPAN13 | |
Recombinant protein corresponding to Human TSPAN13 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only