Learn More
TSPAN2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553355
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VAIRHVQTMY EEAYNDYLKD RGKGNGTLIT FHSTFQCCGK ESSEQVQPTC PKELLGHKNC IDEIETIISV KLQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 89%.
TSPAN2 Polyclonal antibody specifically detects TSPAN2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TSPAN2 | |
Polyclonal | |
Unconjugated | |
TSPAN2 | |
6330415F13Rik; B230119D02Rik; NET3; new EST tetraspan 3; tetraspan 2; tetraspan NET-3; tetraspan TM4SF; tetraspanin 2; tetraspanin 2 isoform; tetraspanin-2; TSN2; TSPAN2; TSPAN-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10100 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60636 | |
TSPAN2 | |
Recombinant protein corresponding to Human TSPAN2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only