Learn More
TSPAN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA553408
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NGTNPDAASR AIDYVQRQLH CCGIHNYSDW ENTDWFKETK NQSVPLSCCR ETASNCNGSL AHPSDLYAEG CEALVVKKLQ EI Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
TSPAN3 Polyclonal antibody specifically detects TSPAN3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TSPAN3 | |
Polyclonal | |
Unconjugated | |
TSPAN3 | |
1700055K04Rik; H41D4 protein; OAP-1; OSP-associated protein 1; tetraspan 3; tetraspan TM4SF; tetraspanin 3; tetraspanin TM4-A; tetraspanin TM4-A homolog; tetraspanin-3; TM4-A; Tm4sf8; Transmembrane 4 superfamily member 8; TSPAN3; TSPAN-3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10099 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O60637 | |
TSPAN3 | |
Recombinant protein corresponding to Human TSPAN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |