missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16887-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Required during maturation of the 40S ribosomal subunit in the nucleolus.Spezifikation
TSR1 | |
Polyclonal | |
Unconjugated | |
TSR1 | |
KIAA1401, TSR1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
100360406, 104662, 55720 | |
-20°C | |
Liquid |
Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q2NL82, Q5SWD9 | |
TSR1 | |
TSR1 Fusion Protein Ag10394 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |