Learn More
TSSC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556683
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: VILSNMVSIS SEPFGHLVDD DDISDQEDHR SEEKSKEPLQ DNVIATYEEH EDSVYAVDWS SADPWLFASL SYDGRLVINR VPRALKYH Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
TSSC1 Polyclonal antibody specifically detects TSSC1 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TSSC1 | |
Polyclonal | |
Unconjugated | |
EIPR1 | |
EARP and GARP complex-interacting protein 1; EIPR1; Endosome-associated recycling protein-interacting protein; Golgi-associated retrograde protein-interacting protein; protein TSSC1; Tssc1; tumor suppressing subtransferable candidate 1; tumor-suppressing STF cDNA 1 protein; Tumor-suppressing subchromosomal transferable fragment candidate gene 1 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7260 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q53HC9 | |
EIPR1 | |
Recombinant protein corresponding to Human TSSC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only