Learn More
TTC1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557715
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: PSIHQAREAC MRLPKQIEER NERLKEEMLG KLKDLGNLVL RPFGLSTENF QIKQDSSTGS YSINFVQNPN NNR Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 97%.
TTC1 Polyclonal antibody specifically detects TTC1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Spezifikation
TTC1 | |
Polyclonal | |
Unconjugated | |
Ttc1 | |
4833412C19Rik; C79328; tetratricopeptide repeat domain 1; tetratricopeptide repeat protein 1; TPR repeat protein 1; TPR1; Ttc1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7265 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q99614 | |
Ttc1 | |
Recombinant protein corresponding to Human TTC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only