Learn More
TTC15 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557063
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AFLHLGQNNF AEAHRFFTEI LRMDPRNAVA NNNAAVCLLY LGKLKDSLRQ LEAMVQQDPR HYLHESVLFN LTTMYELESS RSMQKKQ Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
TTC15 Polyclonal antibody specifically detects TTC15 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spezifikation
TTC15 | |
Polyclonal | |
Unconjugated | |
TRAPPC12 | |
CGI-87; tetratricopeptide repeat domain 15; tetratricopeptide repeat protein 15; TPR repeat protein 15; Trafficking of membranes and mitosis; trafficking protein particle complex 12; trafficking protein particle complex subunit 12; TRAMM; TRAPPC12; TTC15; TTC-15 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51112 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WVT3 | |
TRAPPC12 | |
Recombinant protein corresponding to Human TTC15 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only