Learn More
TTC23L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA558114
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ASPIRIPTVS NDIDWDFCFR MSQQTEIPAH QQTDELYPTG GCGESEEETK AKEKEKAIDC MSHPKEKLAQ SQKKVAQLIK EKMNTQANKE LIR Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 51%.
TTC23L (Tetratricopeptide Repeat Domain 23 Like) is a Protein Coding gene. An important paralog of this gene isTTC23.Spezifikation
TTC23L | |
Polyclonal | |
Unconjugated | |
TTC23L | |
4930401A09Rik; MC25-1; tetratricopeptide repeat domain 23 like; tetratricopeptide repeat domain 23-like; tetratricopeptide repeat protein 23-like; Ttc23l | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
153657 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6PF05 | |
TTC23L | |
Recombinant protein corresponding to Human TTC23L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only