Learn More
TTC6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA564406
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NEYVLMNRAI TNTILKKYEE AKEDFANVIE SCPFWAAVYF NRAHFYYCLK QYELAEEDLN KALSLKPNDA LVYNFRAKVR GKIGLIEEAM ADYNQALDL Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 30%.
TTC6 Polyclonal antibody specifically detects TTC6 in Human samples. It is validated for Immunocytochemistry.Spezifikation
TTC6 | |
Polyclonal | |
Unconjugated | |
TTC6 | |
C14orf25; NCRNA00291; tetratricopeptide repeat domain 6; tetratricopeptide repeat protein 6; TPR repeat protein 6; TTC6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
319089 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86TZ1 | |
TTC6 | |
Recombinant protein corresponding to Human TTC6 | |
100 μL | |
Primary | |
Human | |
Antibody | |