Learn More
TTC7A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA557813
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: ILGKLHYVEG SYRDAISMYA RAGIDDMSME NKPLYQMRLL SEAFVIKGLS LERLPNSIAS RFRLTEREEE VITCFE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
The function of this protein remains unknown.Spezifikation
TTC7A | |
Polyclonal | |
Unconjugated | |
TTC7A | |
1110035E02Rik; 1700007L07Rik; fsn; GIDID; hea; KIAA1140; MINAT; tetratricopeptide repeat domain 7; tetratricopeptide repeat domain 7A; tetratricopeptide repeat protein 7A; TPR repeat protein 7A; Ttc7; TTC7A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57217 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9ULT0 | |
TTC7A | |
Recombinant protein corresponding to Human TTC7A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only