Learn More
TTLL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561731
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: SLTSSTANDR ILKYNLINDT LNIAVPNGEI PDCKWNKSPP KEVLGNYEIL YDEELAQGDG ADRELRSRQG QSLGPRAGRS RDSGRAVLTT WK Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 93%.
Catalytic subunit of the neuronal tubulin polyglutamylase complex. Modifies alpha- and beta-tubulin, generating side chains of glutamate on the gamma-carboxyl groups of specific glutamate residues within the C-terminal tail of alpha- and beta-tubulin.Spezifikation
TTLL1 | |
Polyclonal | |
Unconjugated | |
TTLL1 | |
6330444E16Rik; AV014541; bM121M7.1 (novel protein (ortholog of human tubulin tyrosine ligase-like 1 (TTLL1)), isoform 1); C22orf7; catalytic subunit of neural tubulin polyglutamylase; HS323M22B; p49; PGs3; Probable tubulin polyglutamylase TTLL1; TTLL1; Tubulin polyglutamylase complex subunit 3; tubulin tyrosine ligase like 1; tubulin tyrosine ligase-like 1; tubulin tyrosine ligase-like family member 1; tubulin tyrosine ligase-like family, member 1; tubulin-tyrosine ligase; tubulin--tyrosine ligase-like protein 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25809 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95922 | |
TTLL1 | |
Recombinant protein corresponding to Human TTLL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only