Learn More
TTLL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA562403
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: KLVGTKALST TGKALRTLPT AKVFISLPPN LDFKVAPSIL KPRKAPALLC LRGPQLEVPC CLCPLKSEQF LAPV Highest antigen sequence identity to the following orthologs: Mouse - 46%, Rat - 26%.
TTLL3 Polyclonal antibody specifically detects TTLL3 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TTLL3 | |
Polyclonal | |
Unconjugated | |
TTLL3 | |
HOTTL; PRO0207; TTLL3; Tubulin monoglycylase TTLL3; tubulin tyrosine ligase like 3; tubulin tyrosine ligase-like family member 3; tubulin tyrosine ligase-like family, member 3; tubulin--tyrosine ligase-like protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26140 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4R7 | |
TTLL3 | |
Recombinant protein corresponding to Human TTLL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only