Learn More
TTLL9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA559719
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: GITWIMKPVA RSQGKGIFLF RRLKDIVDWR KDTRSSDDQK DDIPVENYVA QRYIE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 66%.
TTLL9 Polyclonal antibody specifically detects TTLL9 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TTLL9 | |
Polyclonal | |
Unconjugated | |
TTLL9 | |
C20orf125; dJ310O13.1; Probable tubulin polyglutamylase TTLL9; TTLL9; tubulin tyrosine ligase like 9; tubulin tyrosine ligase-like family, member 9; tubulin--tyrosine ligase-like protein 9 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
164395 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3SXZ7 | |
TTLL9 | |
Recombinant protein corresponding to Human TTLL9 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only