Learn More
TTYH1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA554845
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: NSETSDGVSQ LSSALLHANH TLSTIDHLVL ETVERLGEAV RTELTTLEEV LEPRTEQVAE NV Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 85%.
TTYH1 Polyclonal antibody specifically detects TTYH1 in Human samples. It is validated for Immunohistochemistry.Spezifikation
TTYH1 | |
Polyclonal | |
Unconjugated | |
TTYH1 | |
4930459B04Rik; 6330408P11Rik; hTTY1; mTTY1; Protein tweety homolog 1; tty; TTY1; TTYH1; tweety family member 1; tweety homolog 1; tweety homolog 1 (Drosophila) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57348 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H313 | |
TTYH1 | |
Recombinant protein corresponding to Human TTYH1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only