missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBAL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561118
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: IQPNGVVLDT QQDQLENAKM EHTNASFDTF F Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 32%.
Tubulin is the major constituent of microtubules. It binds two moles of GTP, one at an exchangeable site on the beta chain and one at a non-exchangeable site on the alpha chain.Spezifikation
TUBAL3 | |
Polyclonal | |
Unconjugated | |
Tubal3 | |
TUBAL3; Tubulin alpha chain-like 3; tubulin alpha like 3; tubulin, alpha-like 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
79861 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NHL2 | |
Tubal3 | |
Recombinant protein corresponding to Human TUBAL3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only