missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUBB4Q Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA548754
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
TUBB4Q Polyclonal antibody specifically detects TUBB4Q in Human samples. It is validated for Western Blot.Spezifikation
TUBB4Q | |
Polyclonal | |
PBS with 0.09% sodium azide | |
0 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
56604 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
Unconjugated | |
TUBB7P | |
TUBB7P | |
KLH conjugated synthetic peptide between 64-93 amino acids from the N-terminal region of human TUBB4Q. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |