Learn More
Hirschmann Tubing Pharmed BPT Extension Tube ID 0.89mm, rollof 3M
Marke:  Hirschmann 95213503
Tubing Pharmed BPT Extension Tube ID 0.89mm, rollof 3M
Artikelnummer. 17730898
  • ONLINE EXCLUSIVE PRICE
    99.28 €
    64.64 € / 3 Meter
    Sparen 34.64 €
    34% Rabatt