missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TULP4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 55155-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
This antibody is specific to TULP4.
TULP4, also named as KIAA1397, TUBL4 and TUSP, belongs to the TUB family. It may be a substrate-recognition component of a SCF-like ECS (Elongin-Cullin-SOCS-box protein) E3 ubiquitin ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.Spezifikation
TULP4 | |
Polyclonal | |
Unconjugated | |
TULP4 | |
KIAA1397, tubby like protein 4, Tubby related protein 4, Tubby superfamily protein, TUBL4, TULP4, TUSP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
499016, 56995, 68842 | |
-20°C | |
Liquid |
Western Blot | |
0.26 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9JIL5, Q9NRJ4 | |
Tulp4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |