missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TUSC2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA550882
Weitere Details : Netto-Gewicht : 0.01000kg
Beschreibung
The antibody detects endogenous levels of total TUSC2 protein.
TUSC2 Polyclonal antibody specifically detects TUSC2 in Human samples. It is validated for Immunohistochemistry (Paraffin).Spezifikation
TUSC2 | |
Polyclonal | |
Unconjugated | |
TUSC2 | |
1190001E22Rik; AA407686; C3orf11; Fus1; Fus-1 protein; Fusion 1 protein; Lgcc; lung cancer candidiate, FUS; PAP; PDAP2; PDGFA associated protein 2; PDGFA-associated protein 2; Tumor suppressor candidate 2; TUSC 2; Tusc2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11334 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
2.9 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O75896 | |
TUSC2 | |
Full length fusion protein of Human TUSC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |