missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ty1 Tag Monoclonal Antibody (BB2), Invitrogen™
Mouse Monoclonal Antibody
Marke: Invitrogen MA523513
Weitere Details : Netto-Gewicht : 0.00900kg
Beschreibung
Ty1 Tag Monoclonal antibody specifically detects Ty1 Tag in Tag samples. It is validated for ChIP Assay, Western Blot.Spezifikation
Ty1 Tag | |
Monoclonal | |
2.26 mg/mL | |
PBS with 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein A | |
RUO | |
-20°C or -80°C if preferred | |
Liquid | |
Tag |
ChIP Assay, Western Blot | |
BB2 | |
Unconjugated | |
Ty1 Epitope Tag | |
Ty1 tag (amino acid sequence EVHTNQDPLD). | |
50 μg | |
Primary | |
Antibody | |
IgG1 |