Learn More
TYW1B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA561415
Weitere Details : Netto-Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: RVMSRGEGDC DVVKSKHGSI EANFRAWKTK FISQLQALQK GERKKSCGGH CKKGKCESHQ HGSEEREEGS QEQDELHHR Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 67%.
TYW1B Polyclonal antibody specifically detects TYW1B in Human samples. It is validated for Immunohistochemistry.Spezifikation
TYW1B | |
Polyclonal | |
Unconjugated | |
TYW1B | |
LINC00069; long intergenic non-protein coding RNA 69; MGC87315; NCRNA00069; radical S-adenosyl methionine and flavodoxin domain-containing protein 2; radical S-adenosyl methionine and flavodoxin domains 1; RSAFD2; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase; S-adenosyl-L-methionine-dependent tRNA 4-demethylwyosine synthase TYW1B; tRNA wybutosine-synthesizing protein 1 homolog B; tRNA-yW synthesizing protein 1 homolog B; tRNA-yW synthesizing protein 1 homolog B (non-protein coding); TYW1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
441250 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6NUM6 | |
TYW1B | |
Recombinant protein corresponding to Human TYW1B | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only