missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UBE2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 10733-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2B is a member of the E2 ubiquitin-conjugating enzyme family. This enzyme is required for post-replicative DNA damage repair. Its protein sequence is 100% identical to the mouse, rat, and rabbit homologs, which indicates that this enzyme is highly conserved in eukaryotic evolution.Spezifikation
UBE2B | |
Polyclonal | |
Unconjugated | |
UBE2B | |
E2 17kDa, HHR6B, HR6B, RAD6 homolog B, RAD6B, UBC2, UBE2B, Ubiquitin carrier protein B, Ubiquitin protein ligase B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22210, 7320, 81816 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P63146, P63147, P63149 | |
UBE2B | |
UBE2B Fusion Protein Ag1147 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |