missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
N.V.
Spezifikation
Antigen | ULBP-1 |
---|---|
Konzentration | LYOPH |
Verdünnung | Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction |
Anwendungen | Western Blot, Blockade of Receptor-ligand Interaction |
Klassifikation | Polyclonal |
Beschreibung
ULBP-1 Polyclonal specifically detects ULBP-1 in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Spezifikation
ULBP-1 | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
Polyclonal | |
Goat | |
Human | |
ALCAN-beta, MULT-1, N2DL1, N2DL-1, NKG2D ligand 1, NKG2DL, NKG2DL1, RAET1I, RAET1Ialcan-beta, Retinoic acid early transcript 1I, UL16 binding protein 1, UL16-binding protein 1, ULBP1 | |
ULBP1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Blockade of Receptor-ligand Interaction | |
Unconjugated | |
RUO | |
Q9BZM6 | |
80329 | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ULBP-1 Gly26-Pro215 Accession # Q9BZM6 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human ULBP-1 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) ULBP-2 and rhULBP-3 is observed. |
For Research Use Only