Learn More
Use1/UBE2Z Antibody, PE, Novus Biologicals™
Artikelnummer. 18647142 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18647142 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 18647142 Lieferant Novus Biologicals Lieferanten-Nr. NBP297553PE
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Use1/UBE2Z Polyclonal antibody specifically detects Use1/UBE2Z in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen Use1/UBE2Z
Anwendungen Western Blot, Immunocytochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Klassifikation Polyclonal
Konjugat PE
Verdünnung Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin
Zusammensetzung PBS
Gen-Alias MDS032, P31, protein p31, putative MAPK activating protein PM26, Putative MAPK-activating protein PM26, Q-SNARE, SLT1, SNARE-like tail-anchored protein 1 homolog, unconventional SNARE in the ER 1 homolog (S. cerevisiae), USE1L, USE1-like protein, vesicle transport protein USE1
Wirtsspezies Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Use1/UBE2Z fragment.
Reinigungsverfahren Antigen and protein A Affinity-purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Apoptosis, Cell Biology, Cell Cycle and Replication
Primär oder sekundär Primary
Gen-ID (Entrez) 55850
Zielspezies Human
Inhalt und Lagerung Store at 4C in the dark.
Form Purified
Isotype IgG