missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
USP3 Polyclonal specifically detects USP3 in Human samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative).
Spezifikation
Spezifikation
| Antigen | USP3 |
| Anwendungen | Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot |
| Klassifikation | Polyclonal |
| Konzentration | 1.0 mg/mL |
| Konjugat | Unconjugated |
| Verdünnung | Immunohistochemistry 1:1000 - 1:5000, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:1000 - 1:5000, Western Blot (Negative) |
| Zusammensetzung | Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide |
| Gen-Zugriffsnummer | AAD42992.1 |
| Gen-Alias | Deubiquitinating enzyme 3, EC 3.1.2.15, EC 3.4.19.12, MGC129878, MGC129879, SIH003, ubiquitin carboxyl-terminal hydrolase 3, ubiquitin specific peptidase 3, ubiquitin specific protease 3, ubiquitin thioesterase 3, Ubiquitin thiolesterase 3, Ubiquitin-specific-processing protease 3, UBP |
| Gensymbole | USP3 |
| Mehr anzeigen |
For Research Use Only
Name des Produkts
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?