Learn More
ACROBiosystems Vaccinia Virus B18R Protein, His Tag
Marke: ACROBiosystems B1R-H52H6-500UG
Weitere Details : Netto-Gewicht : 0.00001kg
Beschreibung
B18R is a type I interferon (IFN)-binding protein and it is encoded by the B18R open reading frame in the WR (Western Reserve) strain of vaccinia virus, which contains two Ig-like C2-type domains and one Ig-like V-type domain. B18R exists in a soluble and a membrane-bound form. As a type I IFN receptor, B18R counteracts the antiviral effects of host IFN-alpha/beta. Also, B18R acts as a soluble IFN-alpha receptor and thus inhibits the interaction between host IFN-alpha and its receptor.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
40.3 kDa | |
Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession # P25213), was produced in human 293 cells (HEK293) (at ACROBiosystems.) | |
-20°C | |
B18R | |
Vaccinia virus | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |
500 μg | |
His 20 - Glu 351 | |
B18R | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |