missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Vaccinia Virus B18R Protein, His Tag
544.34 € - 2552.37 €
Spezifikation
Molekulargewicht | 40.3 kDa |
---|---|
Quelle | Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession # P25213), was produced in human 293 cells (HEK293) (at ACROBiosystems.) |
Immunogen | His 20 - Glu 351 |
Lagerungsbedingungen | -20°C |
Gen-Alias | B18R |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
15895927
|
ACROBiosystems
B1R-H52H6-500UG |
500 μg |
2552.37 €
500 Mikrogramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
15805937
|
ACROBiosystems
B1R-H52H6-50UG |
50 μg |
544.34 €
50 Mikrogramm |
Verfügbar ab: 03-05-2024 Loggen Sie sich ein, um den Lagerbestand zu sehen |
Bitte melden Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute! | ||||
Beschreibung
B18R is a type I interferon (IFN)-binding protein and it is encoded by the B18R open reading frame in the WR (Western Reserve) strain of vaccinia virus, which contains two Ig-like C2-type domains and one Ig-like V-type domain. B18R exists in a soluble and a membrane-bound form. As a type I IFN receptor, B18R counteracts the antiviral effects of host IFN-alpha/beta. Also, B18R acts as a soluble IFN-alpha receptor and thus inhibits the interaction between host IFN-alpha and its receptor.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 40.3 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spezifikation
40.3 kDa | |
His 20 - Glu 351 | |
B18R | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Vaccinia Virus B18R, His Tag, His 20 - Glu 351 (Accession # P25213), was produced in human 293 cells (HEK293) (at ACROBiosystems.) | |
-20°C | |
B18R | |
Vaccinia virus | |
PBS, pH7.4 | |
HEK293 | |
Viral Proteins |