Learn More
VAMP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 15546-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Involved in the targeting and/or fusion of transport vesicles to their target membrane. Involved for dense-granule secretion in platelets. Plays a role in regulated enzyme secretion in pancreatic acinar cells. Involved in the abscission of the midbody during cell division, which leads to completely separate daughter cells. Involved in the homotypic fusion of early and late endosomes.Spezifikation
VAMP8 | |
Polyclonal | |
Unconjugated | |
VAMP8 | |
EDB, Endobrevin, VAMP 8, VAMP8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22320, 83730, 8673 | |
-20°C | |
Liquid |
Western Blot | |
0.39 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70404, Q9BV40, Q9WUF4 | |
VAMP8 | |
VAMP8 Fusion Protein Ag7903 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |