missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VAP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 14365-1-AP-150UL
Weitere Details : Netto-Gewicht : 0.00010kg
Beschreibung
Copper amine oxidases catalyze the oxidative conversion of amines to aldehydes in the presence of copper and quinone cofactor. The product is a major protein on the adipocyte plasma membrane. It has adhesive properties and also has functional monoamine oxidase activity. A pseudogene for this gene has been described and is located approximately 9-kb downstream.Spezifikation
VAP1 | |
Polyclonal | |
Unconjugated | |
AOC3 | |
AOC3, Copper amine oxidase, HPAO, Membrane primary amine oxidase, SSAO, VAP 1, VAP1, Vascular adhesion protein 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8639 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16853 | |
AOC3 | |
VAP1 Fusion Protein Ag5683 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |