missing translation for 'onlineSavingsMsg'
Learn More

Vasorin/SLIT-like 2 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody has been used in 1 publication

N.V.

Spezifikation

Antigen Vasorin/SLIT-like 2
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Vasorin extracellular domain
Gen-ID (Entrez) 114990
Zusammensetzung Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Rekonstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Mehr Spezifikationen ansehen

Beschreibung

Beschreibung

Vasorin/SLIT-like 2 Polyclonal specifically detects Vasorin/SLIT-like 2 in Human samples. It is validated for Western Blot.
Spezifikation

Spezifikation

Vasorin/SLIT-like 2
114990
Reconstitute at 0.2 mg/mL in sterile PBS.
Goat
IgG
Affinity Purified
RUO
Chinese hamster ovary cell line CHO-derived recombinant human Vasorin extracellular domain
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Primary
Detects human Vasorin in direct ELISAs and Western blots.
Unconjugated
Western Blot
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Produktzertifizierungen

For Research Use Only