Learn More
E2F1(phospho S364) & E2F1 Protein Phosphorylation Antibody Pair, Abnova™
Beschreibung
This protein phosphorylation antibody pair set comes with two antibodies, one against the E2F1 protein, and the other against the specific S364 phosphorylated site of E2F1 for use in in situ Proximity Ligation Assay.
Spezifikation
Spezifikation
| Antigen | E2F1 |
| Anwendungen | In situ PLA |
| Konjugat | Unconjugated |
| Gen | E2F transcription factor 1 |
| Gen-Alias | E2F-1/RBAP1/RBBP3/RBP3 |
| Gensymbole | E2F1 |
| Menge | 1 Set |
| Regulatorischer Status | RUO |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 1869 |
| Mehr anzeigen |