Learn More
Atxn1, Mouse anti-Mouse, ATTO488, Clone: S76-8, Abnova™
Spezifikation
Spezifikation
| Antigen | Atxn1 |
| Anwendungen | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassifikation | Monoclonal |
| Klon | S76-8 |
| Konjugat | Atto 488 |
| Verdünnung | Immunocytochemistry (1:100) Immunofluorescence (1:100) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) Immunoprecipitation Western Blot (1:1000) The optimal working dilution should be determined by the end user. |
| Zusammensetzung | In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). |
| Gen | ataxin 1 |
| Gen-Alias | Ataxin-1/Atx1/C85907/Sca1 |
| Gensymbole | Atxn1 |
| Mehr anzeigen |