missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Vimentin Monoclonal Antibody (VI-01)
GREENER_CHOICE

Artikelnummer. 11584532
Change view
Click to view available options
Menge:
100 μg
Packungsgröße:
100 Mikrogramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
11584532 100 μg 100 Mikrogramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 11584532 Lieferant Invitrogen™ Lieferanten-Nr. MA119168

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Mouse Monoclonal Antibody

This antibody reacts with vimentin, a 57 kDa intermediate filament intracellular protein expressed in variety of mesenchymal and mesodermal cell types. Cross-reactivity was found with smooth muscle desmin. Immunocytochemistry: Staining technique: (a) fix cells for 10 min in methanol at -20°C and for 6 min in acetone at -20°C; (b) fix cells directly in methanol for 10 min at -20°C or in acetone for 10 min at -20°C. Positive control: 3T3 murine Swiss albino fibroblast cell line, RBL rat basophilic leukemia cell line. Flow cytometry: Intracellular staining.

Vimentin is a developmentally regulated intermediate filament protein (IFP) found in cells of mesenchymal origin. It is believed to be involved with the intracellular transport of proteins between the nucleus and plasma membrane. Unlike other IFP proteins, vimentin is expressed, along with desmin, during the early stages of cellular development. During the development process, vimentin is exchanged for new, tissue-specific IFPs. Vimentin has been implicated to be involved in the rate of steroid synthesis via its role as a storage network for steroidogenic cholesterol containing lipid droplets. Vimentin phosphorylation by a protein kinase causes the breakdown of intermediate filaments and activation of an ATP and myosin light chain dependent contractile event. This results in cytoskeletal changes that facilitate the interaction of the lipid droplets within mitochondria, and subsequent transport of cholesterol to the organelles leading to an increase in steroid synthesis.
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen Vimentin
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Monoclonal
Klon VI-01
Konzentration 1 mg/mL
Konjugat Unconjugated
Zusammensetzung TBS with 15mM sodium azide; pH 8
Gen Vim
Gen-Zugriffsnummer P02543, P08670, P20152
Gen-Alias cb28; class-III intermediate microfilament; CTRCT30; epididymis luminal protein 113; FLJ36605; Hel113; intermediate filament; intermediate filament protein; RP11-124N14.1; similar to vimentin; VIM; VIME; Vimentin; vimentin C-terminus
Gensymbole Vim
Wirtsspezies Mouse
Immunogen Pellet of pig brain cold stable proteins after depolymerization of Microtubules.
Reinigungsverfahren Sequential chromatography
Menge 100 μg
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 100522394, 22352, 7431
Zielspezies Human, Mouse, Pig, Mammalia, Rat
Inhalt und Lagerung 4°C, do not freeze
Produkttyp Antibody
Form Liquid
Isotype IgM
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.