Learn More
Vimentin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Artikelnummer. p-7087924 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16806713 20 μL 20 Mikroliter
16896703 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16806713 Lieferant Proteintech Lieferanten-Nr. 103661AP20UL
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
Vimentin is a member of the intermediate filament family. Intermediate filamentents, along with microtubules and actin microfilaments, make up the cytoskeleton. The protein is responsible for maintaining cell shape, integrity of the cytoplasm, and stabilizing cytoskeletal interactions. It is also involved in the immune response, and controls the transport of low-density lipoprotein (LDL)-derived cholesterol from a lysosome to the site of esterification. It functions as an organizer of a number of critical proteins involved in attachment, migration, and cell signaling. Mutations in this gene causes a domit, pulverulent cataract.
Spezifikation
Antigen Vimentin
Anwendungen Western Blot, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry
Klassifikation Polyclonal
Konzentration 0.27 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen VIM
Gen-Zugriffsnummer P08670, P20152, P31000
Gen-Alias VIM, vimentin
Gensymbole VIM
Wirtsspezies Rabbit
Immunogen Vimentin Fusion Protein Ag0489
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 22352, 7431, 81818
Zielspezies Mouse, Human, Rat
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG