missing translation for 'onlineSavingsMsg'
Learn More

Vitronectin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 15833-1-AP-150UL

Weitere Details : Netto-Gewicht : 0.00010kg

 Weitere Artikel aus diesem Sortiment

Artikelnummer. 16885194

  • 329.85 € / 150 Mikroliter

Verfügbar ab
17-06-2021

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!


Beschreibung

Beschreibung

Vitronectin is a plasma glycoprotein that circulates in the blood. Vitronectin is circulating as a mixture of both 75 kDa and 65 kDa forms. Vitronectin is a major cell adhesive glycoprotein and is a common component of extracellular matrix and plasma. It competes effectively with other plasma proteins and is often involved in cell attachment, regulation of blood coagulation and immune responses. It has similar tissue distribution to fibronectin and also its integrin receptor recognises fibronectin.
Spezifikation

Spezifikation

Vitronectin
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P04004, P29788
S protein, Serum spreading factor, V75, vitronectin, VN, VNT, VTN
Rabbit
IgG
150 μL
-20°C
Primary
22370, 29169, 7448
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
VTN
Liquid
Vtn
Vitronectin Fusion Protein Ag8443
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation